Colour hygiene test, HY-RiSE®
1.31200.0001
: Merck
- Description:HY-RiSE® (50 tests)
- Pk:50
The HY-RiSE® kit contains a colour hygiene test strip for manual assessment of the general cleanliness of surfaces, especially in the food and beverage industry. Detects food residues and microorganisms, so-called invisible dirt, which can be hazardous to health. The principle for testing is based on detection of NAD/NADH.
- Much more sensitive than simple visual inspection
- Easy to perform and read
- Clear result within five minutes
- No instrument required, all you need in a pocket format