VWR® PBS Tablets, Buffer, Biotechnology Grade
Leverancier: Avantor
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
VWR®
E404-200TABSEA 174 EUR
E404-200TABS
VWR® PBS Tablets, Buffer, Biotechnology Grade
Buffers Buffers voor elektroforese Wasbuffers
Pre-measured tablets for convenient preparation of 1X PBS solution, pH 7,3 to 7,5.
  • May be sterilised by filtration or autoclave, if desired
  • Does not contain calcium or magnesium
Each tablet, when dissolved in 100 ml of water, prepares 1X PBS solution containing 137 mM sodium chloride, 2,7 mM potassium chloride, and 10 mM phosphate buffer.