Netherlands
Order Entry
Marquis drug test kit
Marquis drug test kit
Catalog # 321761U
Supplier:  VWR Chemicals
undefined
Marquis drug test kit
Catalog # 321761U
Supplier:  VWR Chemicals
Local Article Number: VWRC321761U
Price
274,00 €
Log in for availability
Unit of Measure 1 * 1 Kit
Specifications
  • Description:
    Marquis kit for 40 tests, VWR®
  • Pk:
    1 KIT
Specifications
About this item
This kit comprises materials for 40 tests.
Suitable for testing substances like amphetamines and opiates.
Ordering information: Includes eight packs of five ampoules of 0,2 ml of Marquis, 40 ampoule breakers, and 50 stirrers.