Zinc 99.999% (trace metals basis), powder 40 mesh
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7440-66-6
:
Formula:Zn
MDL Number:MFCD00011291
:65.39 g/mol
Boiling Pt:907 °C (1013 hPa)
Melting Pt:420 °C
Density:7,13 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-175-3
UN:1436
ADR:4.3,II
:GHS02
:GHS09
Specification Test Results
- Appearance:Grey Powder
- Total trace metal impurities:≤0.001 %
Specification Test Results
Product Information
- Catalog #Supplier #Pack typePkAvailabilityPrice
- Supplier Article Number: 367260100Local Article Number: ACRO367260100
- Supplier Article Number: 367260500Local Article Number: ACRO367260500
Frequently Bought Together