Netherlands
Order Entry
Nalgene® Bottles, Wide Neck, HDPE, Round, with Screw Cap
   Sustainable Options Available
Certificates
undefined
Nalgene® Bottles, Wide Neck, HDPE, Round, with Screw Cap
   Sustainable Options Available
Certificates
Specifications
  • Catalog No:
  • NALG312106-0001
  • NALG312106-0016
  • NALG312106-0032
  • NALG312106-0008
  • NALG312106-0004
  • Environmentally Preferable:
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Environmental certification:
  • NAR Certificate of Retirement
  • NAR Certificate of Retirement
  • NAR Certificate of Retirement
  • NAR Certificate of Retirement
  • IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
Specifications
Product Information
  • Catalog #
    Supplier #
    Capacity
    Thread
    Material
    Colour
    Neck I-Ø
    Availability
    Price
  • Wide mouth bottle
    Wide mouth bottle
    NALG312106-0001
    312106-0001
    30 ml
    28-415
    HDPE
    Amber
    21 mm
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 2 200,00 €
    2 200,00 €
    1 * 1.000 ST
       2 200,00 €
    Supplier Article Number: 312106-0001
    Local Article Number: NALG312106-0001
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    NAR Certificate of Retirement
      Low Manufacturing Impact
  • Wide mouth bottle
    Wide mouth bottle
    NALG312106-0004
    312106-0004
    125 ml
    38-415
    HDPE
    Amber
    28 mm
    Log in for availability
    1 * 500 ST
    1 * 500 ST - 1 500,00 €
    1 500,00 €
    1 * 500 ST
       1 500,00 €
    Supplier Article Number: 312106-0004
    Local Article Number: NALG312106-0004
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
      Low Manufacturing Impact
  • Wide mouth bottle
    Wide mouth bottle
    NALG312106-0008
    312106-0008
    250 ml
    43-415
    HDPE
    Amber
    33 mm
    Log in for availability
    1 * 250 ST
    1 * 250 ST - 995,00 €
    995,00 €
    1 * 250 ST
       995,00 €
    Supplier Article Number: 312106-0008
    Local Article Number: NALG312106-0008
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    NAR Certificate of Retirement
      Low Manufacturing Impact
  • Wide mouth bottle
    Wide mouth bottle
    NALG312106-0016
    312106-0016
    500 ml
    53-415
    HDPE
    Amber
    44 mm
    Log in for availability
    1 * 125 ST
    1 * 125 ST - 730,00 €
    730,00 €
    1 * 125 ST
       730,00 €
    Supplier Article Number: 312106-0016
    Local Article Number: NALG312106-0016
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    NAR Certificate of Retirement
      Low Manufacturing Impact
  • Wide mouth bottle
    Wide mouth bottle
    NALG312106-0032
    312106-0032
    1000 ml
    63-415
    HDPE
    Amber
    53 mm
    Log in for availability
    1 * 50 ST
    1 * 50 ST - 471,00 €
    471,00 €
    1 * 50 ST
       471,00 €
    Supplier Article Number: 312106-0032
    Local Article Number: NALG312106-0032
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    NAR Certificate of Retirement
      Low Manufacturing Impact