U heeft gezocht op: Destillatiesets
24  results were found
SearchResultCount:"24"
Sort Results
List view Easy View (new)
Rate These Search Results
Leverancier: Lenz Laborglas GmbH & CO.KG
Omschrijving: Borosilicate glass.
Leverancier: Lenz Laborglas GmbH & CO.KG
Omschrijving: These table top reaction units are used for stirring under normal pressure or vacuum, with possibility of addition of liquids and boiling under reflux...
Leverancier: Lenz Laborglas GmbH & CO.KG
Omschrijving: DURAN®, Borosilicate glass.
Catalogus nummer: (LENZ06265597)
Leverancier: Lenz Laborglas GmbH & CO.KG
Omschrijving: DURAN®, Borosilicate glass.
UOM: 1 * 1 ST
Catalogus nummer: (WILMML1800-700)
Leverancier: SP Industries
Omschrijving: Sublimation apparatus is used for purification by sublimation of quantities of about 0,5 to 500 mg under vacuum.
UOM: 1 * 1 ST
Leverancier: DWK Life Sciences
Omschrijving: Manufactured from WHEATON 33 low extractable borosilicate glass that conforms to USP Type I and ASTM E 438 Type I, Class A requirements.
Catalogus nummer: (201-1418)
Leverancier: DWK Life Sciences
Omschrijving: This apparatus is used for the determination of water in crude oil by distillation.
UOM: 1 * 1 ST
Catalogus nummer: (WHEA377160)
Leverancier: DWK Life Sciences
Omschrijving: Manufactured from WHEATON 33 low extractable borosilicate glass that conforms to USP Type I and ASTM E 438 Type I, Class A requirements.
UOM: 1 * 1 ST
Catalogus nummer: (WILMLG-6655-100)
Leverancier: SP Industries
Omschrijving: This distilling apparatus is used to determine volatile oil in drugs by distillation of a weighed quantity of drug in contact with water.
UOM: 1 * 1 ST
Leverancier: Bohlender
Omschrijving: PTFE/PFA.
Catalogus nummer: (201-0215)
Leverancier: WITEG LABORTECHNIK GMBH
Omschrijving: Borosilicate glass.
UOM: 1 * 1 ST
Leverancier: KLINIPATH
Omschrijving: These solvent recycler recycles xylene, xylene substitutes, alcohol and acetone with 90 - 95% recovery.
Leverancier: SciLabware
Omschrijving: Comprises Markham still body, Liebig condenser and connecting joint clip. When suitable steam is supplied, the steam passes through the sample to prom...
Leverancier: WITEG LABORTECHNIK GMBH
Omschrijving: Borosilicate glass 3.3.
Catalogus nummer: (201-0210)
Leverancier: WITEG LABORTECHNIK GMBH
Omschrijving: Supplementary kit for 171021. Consists of the following components:
UOM: 1 * 1 ST
Catalogus nummer: (201-0209)
Leverancier: WITEG LABORTECHNIK GMBH
Omschrijving: Box with standard cut-out details for half-scale device assembly.
UOM: 1 * 1 EA
Bel voor prijs
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
This product is marked as restricted and can only be purchased by approved Shipping Accounts. If you need further assistance, email VWR Regulatory Department at Regulatory_Affairs@vwr.com
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
Dit artikel is geblokkeerd voor uw organisatie. Neem contact op met uw inkoopafdeling voor meer informatie
Het orginele artikel is niet langer beschikbaar. Het alternatief dat wordt getoond is beschikbaar
Product(en) gemarkeerd met dit symbool worden niet meer verkocht cq verkocht tot het einde van de voorraad. Alternatieven zijn mogelijk beschikbaar door te zoeken met het VWR-catalogusnummer dat hierboven wordt vermeld. Voor meer informatie kunt u contact opnemen met customer service: 020-4808410.
1 - 16 of 24
no targeter for Bottom