Bis(tributyltin)sulphate ≥97%
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
39624.09EA
16.8
EUR
39624.09
39624.18
Bis(tributyltin)sulphate ≥97%
Bis(tributyltin)sulphate
Formule:
C₂₄H₅₄O₄SSn₂ MW: 676,18 g/mol Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00185908 CAS nummer: 26377-04-8 EINECS: 247-634-6 UN: 3146 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...