1-Hexanol
PROOCHE136
:
- Pack type:Glass bottle
- Pk:2 mL
- Seq:2145
- Environmentally Preferable:
: Hexyl alcohol, Pentyl carbinol, 1-Hexyl alcohol, 1-Hydroxy hexane
:
:102.18 g/mol
:13,04715
:156 °C (1013 hPa)
:–45 °C
:0.8322 g/cm³ (20 °C)
:60 °C
:Ambient
:203-852-3
:2282
:3,III
:GHS07
:111-27-3
:H₃C(CH₂)₄CH₂OH
:MFCD00002982