2-Propanol, Supelco®
: Merck
:67-63-0
:
:(CH₃)₂CHOH
:MFCD00011674
:60.1 g/mol
:13,05228
:82 °C (1013 hPa)
:–89 °C
:0.786 g/cm³ (20 °C)
:200-661-7
:1219
:3,II
:M-Clarity™ quality level = MQ100
:12 °C
:01-2119457558-25
:GHS02
:GHS07
:Ambient
- Pack typePk