Houd mijn sessie geopend
Eindigt in 
De sessie is verlopen
Uw sessie is verlopen. Voor uw veiligheid bent u uitgelogd.
Wilt u opnieuw inloggen?
Zakken voor bioprocessing, Standard Flexboy®
Leverancier: Sartorius
Flexboy®
Standard Flexboy® Bioprocessing bags are designed for the preparation, storage and transport of biopharmaceutical solutions, intermediates and final bulk products. They provide a single-use alternative to traditional glass, stainless steel and rigid plastic carboys in a large variety of applications.
  • Sterilised by gamma irradiation
  • Cost effective and reduced risk of cross contamination
  • Flexible and high security of supply
  • Process safety and integrity
  • High sterility assurance level
Order
Expand All / Collapse All
Met TPE-buisjes
Omschrijving Inhoud b×l VPE VWR catalogus nummer Hoeveelheid
Gebruiksklare zakken, standaard 0,15 l 85×205 mm 50 391-0199 Blocked
Gebruiksklare zakken, standaard 1 l 155×299 mm 20 391-0200 Blocked
Gebruiksklare zakken, standaard 3 l 223×381 mm 20 391-0201 Blocked
Gebruiksklare zakken, standaard 5 l 332×376 mm 20 391-0202 Blocked
Gebruiksklare zakken, folie: 0,2 µm 5 l 332×376 mm 20 391-0209 Blocked
Gebruiksklare zakken, folie: 0,2 µm 50 l 580×790 mm 10 391-0213 Blocked
Met TPE-buisjes en Sartopore® 2-gammacapsules
Omschrijving Inhoud b×l VPE VWR catalogus nummer Hoeveelheid
Gebruiksklare zakken, folie: 0,2 µm 1 l 155×299 mm 20 391-0207
Gebruiksklare zakken, folie: 0,2 µm 3 l 223×381 mm 20 391-0208 Blocked
With EVA tubes
Omschrijving Inhoud b×l VPE VWR catalogus nummer Hoeveelheid
Ready to use bags 0,05 l 95×134 mm 20 SARTFFB102603 Blocked
Ready to use bags 0,05 l 95×134 mm 20 SARTFFB102640 Blocked
Ready to use bags 0,15 l 85×205 mm 20 SARPFFB102877
Ready to use bags, standard 0,15 l 85×205 mm 20 SARPFFB102643
Ready to use bags 0,5 l 130×241 mm 20 SARPFFB103398
Ready to use bags, standard 0,5 l 130×241 mm 20 SARPFFB102670
Ready to use bags 1 l 155×299 mm 20 SARPFFB102606
Ready to use bags, standard 1 l 155×299 mm 1 SARTFFB103547 Blocked
Ready to use bags 2 l 59×70 mm 20 SARTFFB102703 Blocked
Ready to use bags, standard 3 l 223×381 mm 20 SARTFFB102704 Blocked
Ready to use bags 5 l 332×376 mm 20 SARTFFB102025 Blocked
Ready to use bags, standard 5 ml 59×70 mm 100 SARTFFB115270 Blocked
Ready to use bags 10 l 300×621 mm 20 SARPFFB101974
Gebruiksklare zakken, standaard 10 l 300×621 mm 20 391-0203 Blocked
Ready to use bags 20 l 431×654 mm 20 SARPFFB101961
Gebruiksklare zakken, standaard 20 l 431×654 mm 10 391-0204 Blocked
Gebruiksklare zakken, standaard 50 l 580×790 mm 10 391-0206 Blocked
Ready to use bags, standard 50 l 580×790 mm 20 SARTFFB102340 Blocked
Blocked Dit artikel is geblokkeerd voor uw organisatie. Neem contact op met uw inkoopafdeling voor meer informatie
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
WRITE A REVIEW
Need help?
  • Whatever you have to say, positive or negative, is important to us.
  • Providing feedback takes only seconds.
  • Your feedback will be made available to the public on the web.
VWR.com will publish your name as vwr.com user.
You can use this name, or edit and save a nickname.
We recommend authors use their first and last initial.
Please confirm the industry you work in
Example: Pharmaceutical
Please confirm your role
Example: Clinical Trial Assistant
Please confirm your location
Example: New York, NY
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Sort all by:
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory. Enter 'X' if unknown for VWR Chemicals. Enter a space if unknown for Merck Chemicals
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Of
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Omschrijving Catalogus nummer Availability Unit Uw prijs Price Per Hoeveelheid
Leveringsvoorwaarden
Artikel 1 Algemeen
1.1 In deze voorwaarden hebben de volgende termen de volgende betekenis:
(a) Voorwaarden: deze algemene voorwaarden;
(b) Leverancier: VWR International B.V., en indien
toepasselijk haar rechtsopvolger;
(c) Afnemer: iedere (rechts) persoon die met Leverancier een overeenkomst sluit of heeft gesloten, of aan wie door of namens Leverancier een aanbod of offerte wordt of is gedaan of gericht, of aan wie of in opdracht
van wie door of namens Leverancier een levering wordt of is gedaan, of in opdracht of ten behoeve van wie door of namens Leverancier een of meer diensten worden of zijn verricht.
1.2 Alle door of namens Leverancier gedane aanbiedingen, uitgebrachte offertes, gesloten overeenkomsten en de uitvoering daarvan, alsmede alle door of namens Leverancier verrichte leveringen en diensten, worden
beheerst door deze Voorwaarden.
1.3 Afwijkingen van deze Voorwaarden zijn alleen van toepassing indien schriftelijk door Leverancier en Afnemer overeengekomen.
1.4 De toepasselijkheid van andere algemene voorwaarden dan deze Voorwaarden wordt uitdrukkelijk uitgesloten, daaronder mede begrepen inkoopvoorwaarden en/of andere algemene voorwaarden van of gehanteerd door
Afnemer. Afnemer aanvaardt de toepasselijkheid van deze Voorwaarden, en de uitsluiting van andere algemene voorwaarden, door aanvaarding van een door of namens Leverancier gedaan aanbod of gedane offerte, door het
aangaan van een overeenkomst met Leverancier, door aanvaarding van een door of namens Leverancier gedane levering, of door aanvaarding van door of namens Leverancier verrichte diensten.
Artikel 2 Offertes
2.1 Iedere aanbieding of offerte door of namens Leverancier is vrijblijvend en bindt Leverancier niet, behalve indien en voor zover door Leverancier schriftelijk uitdrukkelijk anders is vermeld of door partijen schriftelijk anders is
overeengekomen.
2.2 Door of namens Leverancier verstrekte prijslijsten, brochures, catalogi, folders en andere gegevens, ook wanneer in digitale vorm, zijn met de grootst mogelijke zorgvuldigheid samengesteld, maar binden Leverancier
slechts indien en voor zover uitdrukkelijk schriftelijk door Leverancier bevestigd.
2.3 Alle in verband met een aanbieding of offerte verstrekte gegevens en afbeeldingen blijven exclusief eigendom van Leverancier. Het is Afnemer niet toegestaan zonder voorafgaande schriftelijke toestemming van Leverancier
enig materiaal geheel of ten dele te kopiëren of te gebruiken. Het gebruik van dit materiaal dient beperkt te blijven tot eigen gebruik door Afnemer in het kader van de aan Leverancier verstrekte opdracht. Op eerste verzoek
van Leverancier dient al het materiaal terstond aan Leverancier te worden geretourneerd.
2.4 Leverancier is gerechtigd om redelijke kosten verbonden aan een aanbieding of offerte aan Afnemer in rekening te brengen indien geen overeenkomst tot stand komt.
Artikel 3 Totstandkoming en inhoud overeenkomst
3.1 Tussen Leverancier en Afnemer komt een overeenkomst tot stand op het moment van aanvaarding door Leverancier van een opdracht of order van Afnemer, waarbij omvang en inhoud van de overeenkomst volgen uit de aanvaarding
door Leverancier. Een door Afnemer bij Leverancier geplaatste bestelling of opdracht wordt door Leverancier als bindend en onherroepelijk beschouwd op het moment dat deze bestelling of opdracht bij Leverancier is ontvangen
in schriftelijke, elektronische of telefonisch vorm of in enige andere vorm, tenzij de betreffende bestelling of opdracht blijkens haar formulering een voorwaardelijk karakter heeft.
3.2 Leverancier wordt slechts gebonden door een overeenkomst indien deze is aangegaan door een of meer personen die bevoegd zijn Leverancier te binden.
3.3 Gehele of gedeeltelijke wijziging of annulering van een bestelling of opdracht kan enkel plaatsvinden met voorafgaande schriftelijke toestemming van Leverancier en op voorwaarde dat reeds door Leverancier verrichte
werkzaamheden door Afnemer worden vergoed. In geval van wijziging of annulering van een bestelling of opdracht is Afnemer gehouden op eerste verzoek van Leverancier de daaraan voor Leverancier verbonden kosten te voldoen, en
is Leverancier gerechtigd deze kosten bij Afnemer in rekening te brengen alsmede, voor zover van toepassing, de levertijd opnieuw vast te stellen.
3.4 Aanpassing of wijziging van een overeenkomst is slechts mogelijk door middel van een door beide partijen getekend geschrift.
3.5 Voor werkzaamheden of opdrachten waarvoor gezien hun aard en/of omvang geen offerte of opdrachtbevestiging wordt verzonden, geldt dat de overeenkomst tot stand komt op het moment dat door of namens Leverancier met de
uitvoering van de levering wordt begonnen.
3.6 Een overeenkomst tussen partijen wordt aangegaan onder de voorwaarde dat de leveranciers en overige contractspartners van Leverancier hun verplichtingen tijdig en op juiste wijze nakomen.
3.7 Leverancier is gerechtigd bij het aangaan van de overeenkomst, althans voordat van de zijde van Leverancier met de uitvoering van de overeenkomst wordt begonnen dan wel die uitvoering wordt voortgezet, van
Afnemer te verlangen dat zij voldoende zekerheid stelt ter zake van de tijdige voldoening door Afnemer aan haar verplichtingen.
3.8 Leverancier is bevoegd derden in te schakelen ten tehoeve van de uitvoering van de overeenkomst. De kosten hiervan zullen aan Afnemer worden doorberekend.
Artikel 4 Prijzen en kosten
4.1 Iedere prijsopgave door Leverancier is vrijblijvend, tenzij door partijen anders schriftelijk is overeengekomen.
4.2 Door Leverancier op enige wijze vermelde prijzen, zijn steeds exclusief BTW en eventuele additionele kosten, waaronder maar niet beperkt tot transportkosten, kosten ter zake van administratie, e.d., tenzij anders vermeld.
Leverancier is gerechtigd, in geval geleverde goederen een bepaalde door Leverancier vast te stellen factuurwaarde niet overstijgen, administratiekosten in rekening te brengen.
4.3 Op specifiek verzoek van Afnemer, kan worden overeengekomen dat Leverancier aan Afnemer goederen en/of diensten levert die niet tot het vaste assortiment van Leverancier behoren (Third Party Procurement). In dat
geval zullen alle daarmee gemoeide kosten (waaronder maar niet beperkt tot personeelskosten), ongeacht de hoogte ervan, door Leverancier aan Afnemer in rekening worden gebracht.
4.4 Alle eventuele kosten die door de leveranciers van Leverancier worden berekend aan Leverancier, zullen door leverancier worden doorberekend aan Afnemer. Zulks geldt met name ook voor Third Party Procurement goederen.
4.5 Voor de levering van ethanol wordt door Leverancier de vastgestelde accijnstoeslag in rekening gebracht. Indien Afnemer aan Leverancier, vóór het plaatsen van de betreffende opdracht of order, een kopie overhandigt van
een vergunning accijnsvrij betrekken, kan vrij van accijns worden geleverd. Eventuele kosten verbonden aan betreffende documentatie, waaronder maar niet beperkt tot personeelskosten, zullen worden doorberekend door
Leverancier aan Afnemer.
4.6 Voor goederen die geconditioneerd transport behoeven, waaronder maar niet beperkt tot transport op bepaalde temperatuur, zullen door Leverancier aan Afnemer naar redelijkheid extra kosten in rekening worden gebracht.
4.7 Leverancier kan op verzoek van Afnemer ter zake van geleverde goederen aan Afnemer een verklaring verstrekken ten aanzien van de herkomst, samenstelling en/of eigenschappen van de goederen. Leverancier kan
voor de afgifte van een dergelijke verklaring aan Afnemer redelijke kosten in rekening brengen.
4.8 Leverancier kan op verzoek van Afnemer bepaalde diensten verlenen, waaronder, maar niet beperkt tot, installatie van apparatuur, advies, genereren van applicaties en/of afgifte van bepaalde documentatie, e.d.
Voor door Leverancier te leveren diensten kan Leverancier aan Afnemer een redelijke vergoeding in rekening brengen.
Artikel 5 Risico
5.1 Het risico van de goederen die onderwerp zijn van een overeenkomst tussen Leverancier en Afnemer zal op Afnemer overgaan op het moment van aflevering van die goederen bij Afnemer.
5.2 Het uitladen van goederen, alsmede eventuele montage, installatie en inbedrijfstelling daarvan, zijn immer voor rekening zijn van Afnemer.
Artikel 6 Levering
6.1 Behalve indien en voor zover schriftelijk anders overeengekomen, geschieden leveringen (niet franco) aan bedrijf (of huis) van Afnemer of op een andere door Afnemer tijdig opgegeven plaats.
6.2 Behalve indien en voor zover schriftelijk anders overeengekomen, geldt als tijdstip van levering het moment van aankomst van de betreffende goederen bij Afnemer. Zulks geldt eveneens indien Leverancier goederen na
levering moet monteren, installeren en/of in bedrijf stellen.
6.3 Eventuele tekorten, manco’s en beschadigingen dienen door Afnemer binnen 24 uur na levering schriftelijk te worden gemeld aan Leverancier, bij gebreke waarvan de geleverde goederen geacht worden in goede orde
compleet en zonder schade of beschadigingen Afnemer te hebben bereikt.
6.4 Leverancier is gerechtigd om te leveren in gedeelten, welke gedeelten in dat geval afzonderlijk kunnen worden gefactureerd. Alsdan is Afnemer verplicht de afzonderlijke facturen te voldoen overeenkomstig het bepaalde in deze