Houd mijn sessie geopend
Eindigt in 
De sessie is verlopen
Uw sessie is verlopen. Voor uw veiligheid bent u uitgelogd.
Wilt u opnieuw inloggen?
Laboflessen, rond, met schroefdop GL 45
Leverancier: Brand
Borosilicaatglas 3.3, met PP-schroefdop.
  • Zeer goede chemische bestendigheid
  • Hoge temperatuurbestendigheid
  • Relatief hoge bestendigheid tegen temperatuurverandering door minimale thermische expansie
Rond, met schaalverdeling, DIN GL 45 schroefdraad, gietring en blauwe PP-schroefdop.
*Met speciaal gevormde glazen rand voor verbeterd uitgieten; geen extra uitgietring nodig
Certificaten: ISO 4796
Order
Expand All / Collapse All
Inhoud Schroefdraad Ø uitw. Hoogte VPE VWR catalogus nummer Hoeveelheid
10000 ml GL 45 234 mm 410 mm 1 BRND122574
1000 ml GL 45 101 mm 225 mm 10 BRND122562
100 ml GL 45 56 mm 100 mm 10 BRND122538
20000 ml GL 45 300 mm 505 mm 1 BRND122578
2000 ml GL 45 136 mm 262 mm 10 BRND122564
25* ml GL 25 36 mm 70 mm 10 BRND122520
250 ml GL 45 70 mm 138 mm 10 BRND122548
5000 ml GL 45 186 mm 335 mm 1 BRND122570
500 ml GL 45 86 mm 176 mm 10 BRND122554
50 ml GL 32 46 mm 87 mm 10 BRND122528
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Accessoires
Expand All / Collapse All
Spare Parts
Omschrijving Schroefdraad VPE VWR catalogus nummer Hoeveelheid
Gietringen GL 32 10 BRND122662
Gietringen GL 45 10 BRND122664
Schroefdoppen GL 25 10 BRND122620
Schroefdoppen GL 32 10 BRND122628
Schroefdoppen GL 45 10 BRND122638
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
WRITE A REVIEW
Need help?
  • Whatever you have to say, positive or negative, is important to us.
  • Providing feedback takes only seconds.
  • Your feedback will be made available to the public on the web.
VWR.com will publish your name as vwr.com user.
You can use this name, or edit and save a nickname.
We recommend authors use their first and last initial.
Please confirm the industry you work in
Example: Pharmaceutical
Please confirm your role
Example: Clinical Trial Assistant
Please confirm your location
Example: New York, NY
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Sort all by:
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory. Enter 'X' if unknown for VWR Chemicals. Enter a space if unknown for Merck Chemicals
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Of
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Omschrijving Catalogus nummer Availability Unit Uw prijs Price Per Hoeveelheid