

Ethanol absolute, dehydrated (max. 0.01% H₂O), SeccoSolv®, Sigma-Aldrich®
Catalog # MERC1.00990.1001_T
Supplier: Merck
CAS Number:
Discontinued
Product has been discontinued. However we found similar alternative products.
Alternatives
Ethanol dried (max. 0.01 % H2O) SeccoSolv® 1 * 500 mL Catalog #1.00990.0500
Product 1.00990.0500 is a replacement for MERC1.00990.1001_T but varies by Brand, Packaging or Quantity.
Specifications
- Pack type:Glass bottle
- Pk:1 L
- Seq:5037
- Duty status:Duty paid
Specifications
Specification Test Results
- Solubility:(20 °C) completely miscible
- Melting Point:-114.5 °C
- Molar Mass:46.07 g/mol
- Boiling Point:78.3 °C (1013 hPa)
- Vapor Pressure:59 hPa (20 °C)
- Flash Point:12 °C
- Refractive Index:1.36
- Explosion Limit:3.5 - 15 %(V)
- Density:0.790 - 0.793 g/cm3 (20 °C)
- pH:7.0 (10 g/l, H2O, 20 °C)
- Ignition Point:363 °C
- Hazard Class:3
- Hazard Class Description:Flammable liquid
Specification Test Results
About this item
:
:46.07 g/mol
Merck Index:12,03806
Boiling Pt:78.3 °C (1013 hPa)
Melting Pt:–117 °C
Density:0.7895 g/cm³ (20 °C)
Flash Pt:12 °C
Storage Temperature:Ambient
EINECS:200-578-6
UN:1170
ADR:3,II
REACH:01-2119457610-43
:GHS02
:GHS07
CAS Number:64-17-5
Formula:C₂H₅OH
MDL Number:MFCD00003568