Diatomaceous earth filter papers, grade 470
Supplier: Sartorius
Certificates
About this item
Grade 470 papers are made of cellulose and diatomaceous earth and offer a much better separating capability than pure cellulose papers at the same rate of filtration.
- Quickly retain the finest particles at high flow rates
Specifications
- Catalog No:
- SARTFT-4-606-320
- SARTFT-4-606-240
- SARTFT-4-606-185
- SARTFT-4-606-150
- SARTFT-4-606-125
- Weight:
- 140 g/m²
- 140 g/m²
- 140 g/m²
- 140 g/m²
- 140 g/m²
- Thickness:
- 0,32 mm
- 0,32 mm
- 0,32 mm
- 0,32 mm
- 0,32 mm
- Filtration speed:
- 80 s
- 80 s
- 80 s
- 80 s
- 80 s
Specifications
Product Information
- Catalog #Supplier #GradeSizePkAvailabilityPrice
- Specifications:
More
Specifications
Environmentally PreferableWeight140 g/m²Thickness0,32 mmFiltration speed80 s - Specifications:
More
Specifications
Environmentally PreferableWeight140 g/m²Thickness0,32 mmFiltration speed80 s - Specifications:
More
Specifications
Environmentally PreferableWeight140 g/m²Thickness0,32 mmFiltration speed80 s - Specifications:
More
Specifications
Environmentally PreferableWeight140 g/m²Thickness0,32 mmFiltration speed80 s - Specifications:
More
Specifications
Environmentally PreferableWeight140 g/m²Thickness0,32 mmFiltration speed80 s