Vacuum stopcocks
Leverancier: Lenz
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
LENZ02901402EA 48.7 EUR
LENZ02901402 LENZ02902408 189-2102 189-2113 189-2124 189-2125 189-2114 189-2103 189-2115 189-2105 189-2122 189-2111 189-2123 189-2112 LENZ02921402 LENZ02961804 LENZ02962408 LENZ02962106 LENZ02961402 LENZ02962910
Vacuum stopcocks
Afsluitkranen
Vacuum stopcocks, DURAN® with side arms and hollow NS glass plug.
  • Excellent chemical resistance
  • Excellent temperature resistance
Leakage rate according to DIN 12540-1.