Vigreux-kolommen met vacuümmantel
Leverancier: Lenz
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
LENZ05136201EA 183 EUR
LENZ05136201 LENZ05136303 LENZ05136304 LENZ05136504
Vigreux-kolommen met vacuümmantel
Onderdelen voor destillatie Destillatiekolommen
DURAN®, borosilicate glass.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
With NS socket, NS cone and with fixed vaccum jacket.
With NS socket, NS cone and with fixed vaccum jacket.