Netherlands
Order Entry
Tissue cassettes in QuickLoad™ sleeves, Slimsette™, M509SL
Supplier:  Simport Scientific
Certificates
undefined
Tissue cassettes in QuickLoad™ sleeves, Slimsette™, M509SL
Supplier:  Simport Scientific
Related Information
Product Information
  • Catalogus #
    Leverancier #
    Colour
    Pk
    Availability
    Price
  • 720-1423
    M509-11SL
    Orange
    750
    Log in for availability
    1 * 750 ST
    1 * 750 ST - 284,00 €
    284,00 €
    1 * 750 ST
       284,00 €
    Supplier Article Number: M509-11SL
    Local Article Number: SIMPM509-11SL
    Specifications
    Environmentally Preferable
  • 720-1432
    M509-3SL
    Pink
    750
    Log in for availability
    1 * 750 ST
    1 * 750 ST - 284,00 €
    284,00 €
    1 * 750 ST
       284,00 €
    Supplier Article Number: M509-3SL
    Local Article Number: SIMPM509-3SL
    Specifications
    Environmentally Preferable
  • 720-1421
    M509-10SL
    Lilac
    750
    Log in for availability
    1 * 750 ST
    1 * 750 ST - 284,00 €
    284,00 €
    1 * 750 ST
       284,00 €
    Supplier Article Number: M509-10SL
    Local Article Number: SIMPM509-10SL
    Specifications
    Environmentally Preferable
  • 720-1430
    M509-2SL
    White
    750
    Log in for availability
    1 * 750 ST
    1 * 750 ST - 284,00 €
    284,00 €
    1 * 750 ST
       284,00 €
    Supplier Article Number: M509-2SL
    Local Article Number: SIMPM509-2SL
    Specifications
    Environmentally Preferable
  • 720-1444
    M509-9SL
    Grey
    750
    Log in for availability
    1 * 750 ST
    1 * 750 ST - 284,00 €
    284,00 €
    1 * 750 ST
       284,00 €
    Supplier Article Number: M509-9SL
    Local Article Number: SIMPM509-9SL
    Specifications
    Environmentally Preferable
  • 720-1442
    M509-8SL
    Tan
    750
    Log in for availability
    1 * 750 ST
    1 * 750 ST - 284,00 €
    284,00 €
    1 * 750 ST
       284,00 €
    Supplier Article Number: M509-8SL
    Local Article Number: SIMPM509-8SL
    Specifications
    Environmentally Preferable
  • 720-1440
    M509-7SL
    Pale orange
    750
    Log in for availability
    1 * 750 ST
    1 * 750 ST - 284,00 €
    284,00 €
    1 * 750 ST
       284,00 €
    Supplier Article Number: M509-7SL
    Local Article Number: SIMPM509-7SL
    Specifications
    Environmentally Preferable
  • 720-1438
    M509-6SL
    Blue
    750
    Log in for availability
    1 * 750 ST
    1 * 750 ST - 284,00 €
    284,00 €
    1 * 750 ST
       284,00 €
    Supplier Article Number: M509-6SL
    Local Article Number: SIMPM509-6SL
    Specifications
    Environmentally Preferable
  • 720-1436
    M509-5SL
    Yellow
    750
    Log in for availability
    1 * 750 ST
    1 * 750 ST - 284,00 €
    284,00 €
    1 * 750 ST
       284,00 €
    Supplier Article Number: M509-5SL
    Local Article Number: SIMPM509-5SL
    Specifications
    Environmentally Preferable
  • 720-1425
    M509-12SL
    Turquoise
    750
    Log in for availability
    1 * 750 ST
    1 * 750 ST - 284,00 €
    284,00 €
    1 * 750 ST
       284,00 €
    Supplier Article Number: M509-12SL
    Local Article Number: SIMPM509-12SL
    Specifications
    Environmentally Preferable
  • 720-1434
    M509-4SL
    Green
    750
    Log in for availability
    1 * 750 ST
    1 * 750 ST - 284,00 €
    284,00 €
    1 * 750 ST
       284,00 €
    Supplier Article Number: M509-4SL
    Local Article Number: SIMPM509-4SL
    Specifications
    Environmentally Preferable