Netherlands
Order Entry
Benzoic acid
Irritant
Benzoic acid
Catalog # PROONIST-39J
CAS Number:  
undefined
Irritant
Benzoic acid
Catalog # PROONIST-39J
CAS Number:  
Supplier Article Number: NIST-39J
Local Article Number: PROONIST-39J
Price
Inquire For Price
Log in for availability
Unit of Measure 1 * 30 g
Specifications
  • Pack type:
    Glass bottle for solids
  • Pk:
    30 g
  • Seq:
    3332
Specifications
About this item
 :
 :122.12 g/mol
Merck Index:13,01092
Boiling Pt:249 °C (1013 hPa)
Melting Pt:121…123 °C
Density:1.316 g/cm³ (20 °C)
Flash Pt:121 °C (closed cup)
Storage Temperature:Ambient
EINECS:200-618-2
REACH:01-2119455536-33
 :GHS07
CAS Number:65-85-0
Formula:C₇H₆O₂
MDL Number:MFCD00002398