Filterkroezen
Leverancier: DWK Life Sciences
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
SCOT258512101EA 186.8 EUR
SCOT258512101 SCOT258512401 SCOT258513303 SCOT258510409 SCOT258511302 SCOT258511105 SCOT258510203 SCOT258512204 SCOT258513106 SCOT258512504 SCOT258513406 SCOT258511208 SCOT258511405 SCOT258510306 SCOT258512307 SCOT258513209 SCOT258513509
Filterkroezen
Kroezen
These filtration crucibles are made from clear DURAN® borosilicate glass 3.3.
  • Good thermal shock and chemical resistance
Typical applications include qualitative inorganic analysis and preparative chemistry.