Ubbelohde viscometers (DIN) with TC sensors
Leverancier: SI Analytics
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
SCOI285423580EA 1250 EUR
SCOI285423580 SCOI285423460 SCOI285423680 SCOI285423560 SCOI285423510 SCOI285423610 SCOI285423550 SCOI285423430 SCOI285423650 SCOI285423530 SCOI285423630 SCOI285423410 SCOI285423590 SCOI285423690 SCOI285423570 SCOI285423450 SCOI285423670 SCOI285423490 SCOI285423620 SCOI285423500 SCOI285423600 SCOI285423700 SCOI285423660 SCOI285423540 SCOI285423420 SCOI285423640 SCOI285423520
Ubbelohde viscometers (DIN) with TC sensors
Ubbelohde viscometers especially designed for liquids that can not be detected with other systems.
  • Calibrated with constant for automatic measurement
  • Opaque, black and/or electrically conductive samples
  • Meniscus passage detected due to the different conductivity of the liquid and gas phases
Measuring levels are marked by TC sensors.
Certificaten: DIN 51562-1, ISO 3105