Netherlands
Order Entry
Petri dishes, sterile
Petri dishes, sterile
Supplier:  Simport Scientific
Certificates
undefined
Petri dishes, sterile
Supplier:  Simport Scientific
Certificates
Related Information
Product Information
  • Catalogus #
    Leverancier #
    Version
    Ø
    Height
    Sterility
    Packed
    Availability
    Price
  • Petri dish, 9 ml
    Petri dish, 9 ml
    SIMPD210-15
    D210-15
    With three vents
    35 mm
    10 mm
    Sterile
    25 bags with 20 pieces
    Log in for availability
    Log in for availability
    1 * 500 ST
    1 * 500 ST - 219,00 €
    219,00 €
    1 * 500 ST
       219,00 €
    Supplier Article Number: D210-15
    Local Article Number: SIMPD210-15
    Specifications:
    Specifications
    Environmentally Preferable
  • Petri dish, 12 ml
    Petri dish, 12 ml
    SIMPD210-14
    D210-14
    Without vents
    50 mm
    9 mm
    Sterile
    25 bags with 20 pieces
    Log in for availability
    Log in for availability
    1 * 500 ST
    1 * 500 ST - 235,00 €
    235,00 €
    1 * 500 ST
       235,00 €
    Supplier Article Number: D210-14
    Local Article Number: SIMPD210-14
    Specifications:
    Specifications
    Environmentally Preferable
  • Petri dish, 28 ml
    Petri dish, 28 ml
    SIMPD210-13
    D210-13
    With three vents
    55 mm
    13 mm
    Sterile
    25 bags with 20 pieces
    Log in for availability
    Log in for availability
    1 * 500 ST
    1 * 500 ST - 248,00 €
    248,00 €
    1 * 500 ST
       248,00 €
    Supplier Article Number: D210-13
    Local Article Number: SIMPD210-13
    Specifications:
    Specifications
    Environmentally Preferable
  • Petri dish, 100 ml
    Petri dish, 100 ml
    SIMPD210-7
    D210-7
    Without vents
    90 mm
    20 mm
    Sterile
    25 bags with 20 pieces
    Log in for availability
    Log in for availability
    1 * 500 ST
    1 * 500 ST - 436,00 €
    436,00 €
    1 * 500 ST
       436,00 €
    Supplier Article Number: D210-7
    Local Article Number: SIMPD210-7
    Specifications:
    Specifications
    Environmentally Preferable
  • Petri dish, 100 ml
    Petri dish, 100 ml
    SIMPD210-7WL
    D210-7WL
    With three vents
    90 mm
    20 mm
    Sterile
    25 bags with 20 pieces
    Log in for availability
    Log in for availability
    1 * 500 ST
    1 * 500 ST - 407,00 €
    407,00 €
    1 * 500 ST
       407,00 €
    Supplier Article Number: D210-7WL
    Local Article Number: SIMPD210-7WL
    Specifications:
    Specifications
    Environmentally Preferable
  • Petri dish, 125 ml
    Petri dish, 125 ml
    SIMPD210-8
    D210-8
    With three vents
    90 mm
    25 mm
    Sterile
    25 bags with 20 pieces
    Log in for availability
    Log in for availability
    1 * 500 ST
    1 * 500 ST - 463,00 €
    463,00 €
    1 * 500 ST
       463,00 €
    Supplier Article Number: D210-8
    Local Article Number: SIMPD210-8
    Specifications:
    Specifications
    Environmentally Preferable
  • Petri dish, 125 ml
    Petri dish, 125 ml
    SIMPD210-8R
    D210-8R
    Without vents
    90 mm
    25 mm
    Sterile
    25 bags with 20 pieces
    Log in for availability
    Log in for availability
    1 * 500 ST
    1 * 500 ST - 512,00 €
    512,00 €
    1 * 500 ST
       512,00 €
    Supplier Article Number: D210-8R
    Local Article Number: SIMPD210-8R
    Specifications:
    Specifications
    Environmentally Preferable