Volumetrische lepels
Leverancier: Bürkle
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
231-2566EA 8.3 EUR
BURK5334-0001
Volumetrische lepels
Disposable spoons made of blue PS.
  • Round shape, easy emptying
  • Capacity is printed on the spoon handle
  • Available as a set with 8 volumetric spoons or separate in bags of 100 volumetric spoons
* set van acht lepels. Van elke maat één.