Microtome, RM 2245
Leverancier: Leica
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
720-1881
Microtome, RM 2245
Microtomen Microtomen
The manual microtome with high-precision motorised specimen feed. Operator fatigue is reduced through the use of the patented spring system, which eliminates the need for a heavy counter balance weight in the hand wheel. Enhanced user comfort results in maximum workplace efficiency and high specimen throughput.
  • Two manual modes: Conventional full hand wheel rotation and rocking mode
  • Force compensation system for extremely smooth manual hand wheel operation
  • Separate and intuitive control panel for instrument settings and communication display integrated in the instrument housing at eye level
  • Programmable specimen retraction, efficient operation with maximum section reproducibility
  • Knife holder concept for disposable blades, with finger guard in contrasting colour