Netherlands
Order Entry
Plasma collection tube with heparin, BD Vacutainer®
Plasma collection tube with heparin, BD Vacutainer®
Supplier:  BD Medical
Certificates
Plasma collection tube with heparin, BD Vacutainer®
Supplier:  BD Medical
Certificates
Related Information
Product Information
  • Catalog #
    Supplier #
    Material
    Closure type
    Additive
    Length
    Ø
    Cap colour
    Availability
    Price
  • Blood Collection tube, BD Vacutainer®
    Blood Collection tube, BD Vacutainer®
    BDAM367876
    367876
    Plastic
    BD Hemogard™
    Sodium heparin (102 IU)
    100 mm
    13 mm
    Green
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 92,70 €
    92,70 €
    1 * 100 ST
       92,70 €
    Supplier Article Number: 367876
    Local Article Number: BDAM367876
    Specifications:
    Specifications
    Environmentally Preferable
  • Blood Collection tube, BD Vacutainer®
    Blood Collection tube, BD Vacutainer®
    BDAM368884
    368884
    PET
    BD Hemogard™
    Lithium heparin
    75 mm
    13 mm
    Green
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 70,30 €
    70,30 €
    1 * 100 ST
       70,30 €
    Supplier Article Number: 368884
    Local Article Number: BDAM368884
    Specifications:
    Specifications
    Environmentally Preferable
  • Blood Collection tube, BD Vacutainer®
    Blood Collection tube, BD Vacutainer®
    BDAM367526
    367526
    PET
    BD Hemogard™
    Lithium heparin (171 IU)
    100 mm
    16 mm
    Green
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 90,00 €
    90,00 €
    1 * 100 ST
       90,00 €
    Supplier Article Number: 367526
    Local Article Number: BDAM367526
    Specifications:
    Specifications
    Environmentally Preferable
  • Blood Collection tube, BD Vacutainer®
    Blood Collection tube, BD Vacutainer®
    BDAM368495
    368495
    Plastic
    BD Hemogard™
    Lithium heparin
    75 mm
    13 mm
    Green
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 271,00 €
    271,00 €
    1 * 1.000 ST
       271,00 €
    Supplier Article Number: 368495
    Local Article Number: BDAM368495
    Specifications:
    Specifications
    Environmentally Preferable
  • Blood Collection tube, BD Vacutainer®
    Blood Collection tube, BD Vacutainer®
    BDAM368480
    368480
    Glass
    BD Hemogard™
    Sodium heparin
    100 mm
    16 mm
    Green
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 200,00 €
    200,00 €
    1 * 100 ST
       200,00 €
    Supplier Article Number: 368480
    Local Article Number: BDAM368480
    Specifications:
    Specifications
    Environmentally Preferable
  • Blood Collection tube, BD Vacutainer®
    Blood Collection tube, BD Vacutainer®
    BDAM368886
    368886
    PET
    BD Hemogard™
    Lithium heparin
    100 mm
    13 mm
    Green
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 622,00 €
    622,00 €
    1 * 1.000 ST
       622,00 €
    Supplier Article Number: 368886
    Local Article Number: BDAM368886
    Specifications:
    Specifications
    Environmentally Preferable
  • Blood Collection tube, BD Vacutainer®
    Blood Collection tube, BD Vacutainer®
    BDAM367869
    367869
    PET
    BD Hemogard™
    Sodium heparin
    75 mm
    13 mm
    Green
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 78,90 €
    78,90 €
    1 * 100 ST
       78,90 €
    Supplier Article Number: 367869
    Local Article Number: BDAM367869
    Specifications:
    Specifications
    Environmentally Preferable