Double-diaphragm pumps, oil-free
Leverancier: Bürkle
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
BURK5765-0004EA 1020 EUR
BURK5765-0004 BURK5765-0002 BURK5765-0003
Double-diaphragm pumps, oil-free
Pneumatic double-diaphragm pump are used to pump viscous, corrosive, abrasive and flammable liquids.
  • Self-priming
  • Maximum temperature 66 °C
Operating compressed air minimum 1,4 bar, maximum 6,8 bar.