VWR® Smoothie™ Pipette Filler
Leverancier: Jencons
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
612-3862EA 25.8 EUR
612-3862 JENC475-373 JENC475-371 612-3864 612-3863 JENC475-372 612-3865
VWR® Smoothie™ Pipette Filler
Pipetten Pipetvullers
The Smoothie™ pipette filler is a simple manual pipetting aid for convenient single-handed operations. It fits all sizes of glass or plastic pipettes from 1 to 25 ml.
  • Comfortable and simple to use
  • Smooth, controllable thumb action
  • Silicone adapter for leakproof positioning of each pipette
The unique range control allows selection from two dispense resolutions. One range setting delivers fast filling with optimal accuracy when used with 10 to 25 ml pipettes, the other setting is for use with pipettes from 1 to 10 ml where accuracy of fill can sometimes be difficult to control.
Opgelet: *De product(en) op deze pagina vallen buiten het toepassingsgebied van de EU-richtlijn 2019/904