Automatic burettes, Schilling pattern, Schellbach stripe, LaboPlast®
Leverancier: Bürkle
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
LaboPlast®
BURK9695-1025EA 60.8 EUR
BURK9695-1025 BURK9695-1015 BURK9695-1005 BURK9695-1050 BURK9695-1010
Automatic burettes, Schilling pattern, Schellbach stripe, LaboPlast®
Buretten
Borosilicate glass 3.3, class B.
  • Adjusted for Ex
  • Blue ring graduations, Schellbach stripe
  • Precise dosing using the press button, exact fine titration with the micro-screw and precise discharge jet
Graduation according to Category B (Tolerances according to DIN EN ISO 385).
Certificaten: Graduation according to Category B (Tolerances according to DIN EN ISO 385).