2,4-Di-tert-butylphenol 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:96-76-4
:
Formula:C₁₄H₂₂O
MDL Number:MFCD00008828
:206.33 g/mol
Boiling Pt:264 °C (760 mmHg)
Melting Pt:54…59 °C
Density:0,887 g/cm³ (20 °C)
Flash Pt:115 °C
Storage Temperature:Ambient
EINECS:202-532-0
UN:3077
ADR:9,III
:GHS07
:GHS09
Specification Test Results
- Appearance:White to yellow Crystalline solid
- Infrared spectrum:Conforms
- Melting point:54 to 59 °C
- Water:≤0.1 % (K.F.)
- GC:≥96.0 %
Specification Test Results
Product Information
- Catalog #Supplier #Pack typePkAvailabilityPrice
- Supplier Article Number: 160121000Local Article Number: ACRO160121000
- Supplier Article Number: 160125000Local Article Number: ACRO160125000
- Supplier Article Number: 160120025Local Article Number: ACRO160120025