Kegs, wide neck, UN approved, with screw lid
Leverancier: Bürkle
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
216-4596EA 30.6 EUR
216-4596 BURK1105-0010 BURK1105-0033 216-5385 BURK1105-0026
Kegs, wide neck, UN approved, with screw lid
Vaten, vaten, trommels
HDPE white containers with red screw lid.
  • Temperature resistant from −20 to + 80°C
  • Cap has integrated rubber gasket to ensure an airtight and watertight seal
  • UN approved
The wide-necked container is approved for bulk goods. Used for disposal or storage. For liquids, powders or granules. Can be closed tightly and sealed.