U heeft gezocht op: Cytologische trechters
10  results were found
SearchResultCount:"10"
Sort Results
List view Easy View (new)
Rate These Search Results
Catalogus nummer: (720-1999)
Leverancier: VWR Collection
Omschrijving: Plastic kamer voor eenmalig gebruik, gebruikt bij cytocentrifugatie voor urine, gynaecologische en niet-gynaecologische monsters, ook ontworpen voor h...
UOM: 1 * 600 ST
Leverancier: Simport Scientific
Omschrijving: The CytoSepâ„¢ funnel chambers for the Hettich Cyto-System optimises laboratory throughput with multiple funnel options in 2, 3, or 4 funnels per slide.
Catalogus nummer: (720-1868)
Leverancier: Simport Scientific
Omschrijving: Cytology funnel for SAKURA Cyto-Tek® centrifuge.
UOM: 1 * 200 ST
Leverancier: Simport Scientific
Omschrijving: These one-way cytology funnels / cytofunnels are used for applying a thin cell layer to a microscope slide by means of centrifugation. The application...
Sale
Leverancier: Simport Scientific
Omschrijving: These funnels will snap quickly in place, allowing the pad to align correctly with the sample tunnel. They are made with a compression ring around the...
Catalogus nummer: (720-1972)
Leverancier: Simport Scientific
Omschrijving: CytoSep™ cytology funnels for the StatSpin Cytofuge® 2 cytocentrifuge allow cells to gently concentrate in a 7 mm diameter area on the slide, while su...
UOM: 1 * 48 ST
Catalogus nummer: (720-1867)
Leverancier: Simport Scientific
Omschrijving: The CytoSepâ„¢ MEGA funnel provides an easy, efficient and cost effective method of producing high quality thin layer slide preparations. It eliminates ...
UOM: 1 * 480 ST
Leverancier: THARMAC GmbH
Omschrijving: Enkel geschikt met EASY rotors JC105 en JC375.
Leverancier: Epredia
Omschrijving: These disposable funnels are used for the shandon Cytofunnel® sample chambers.
Leverancier: Epredia
Omschrijving: A disposable sample chamber with cap and pre-attached white filter card for high absorbency, this product is meant for single use with all cytocentrif...
Bel voor prijs
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
This product is marked as restricted and can only be purchased by approved Shipping Accounts. If you need further assistance, email VWR Regulatory Department at Regulatory_Affairs@vwr.com
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
Dit artikel is geblokkeerd voor uw organisatie. Neem contact op met uw inkoopafdeling voor meer informatie
Het orginele artikel is niet langer beschikbaar. Het alternatief dat wordt getoond is beschikbaar
Product(en) gemarkeerd met dit symbool worden niet meer verkocht cq verkocht tot het einde van de voorraad. Alternatieven zijn mogelijk beschikbaar door te zoeken met het VWR-catalogusnummer dat hierboven wordt vermeld. Voor meer informatie kunt u contact opnemen met customer service: 020-4808410.
1 - 10 of 10
no targeter for Bottom