Houd mijn sessie geopend
Eindigt in 
De sessie is verlopen
Uw sessie is verlopen. Voor uw veiligheid bent u uitgelogd.
Wilt u opnieuw inloggen?
  • Criteria
  • Leverancier
  • Refine by Suppliers
    Sort by:

  • Zoek binnen resultaten

p-Xyleen

SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Click OK to update your results.
 
 
SearchResultCount:"12"
  List view Grid view Easy View (new)
Sorteer op:
 
 
 
 

Leverancier:  Merck
Omschrijving:   p-Xyleen for synthesis
Leverancier:  Merck
Omschrijving:   p-Xyleen, EMSURE® ISO analytical reagent
Leverancier:  Merck
Omschrijving:   p-Xyleen reference substance for gas chromatography
UOM:  1 * 5 ml
Catalogus nummer: (28984.292)
Leverancier:  VWR Chemicals
Omschrijving:   p-Xyleen ≥99%, GPR RECTAPUR®
UOM:  1 * 1 l
Leverancier:  Alfa Aesar
Omschrijving:   p-Xyleen ≥99%
Leverancier:  Acros Organics
Omschrijving:   p-Xyleen ≥99%, Extra Pure
Leverancier:  Acros Organics
Omschrijving:   p-Xyleen ≥99%, pure
Leverancier:  Acros Organics
Omschrijving:   p-Xyleen ≥99% for HPLC
Call for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
This product is marked as restricted and can only be purchased by approved Shipping Accounts. If you need further assistance, email VWR Regulatory Department at Regulatory_Affairs@vwr.com
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
Dit artikel is geblokkeerd voor uw organisatie. Neem contact op met uw inkoopafdeling voor meer informatie
Het orginele artikel is niet langer beschikbaar. Het alternatief dat wordt getoond is beschikbaar
Dit artikel is vervallen en leverbaar zolang de voorraad strekt. Binnen de VWR catalogus kunt u zoeken op alternatieven. Voor meer informatie kunt u contact opnemen met customer service: 020-4808410.
1 - 8  Of 12
  1  2  Volgende

A new look and a renewed commitment

Built on the features & resources you know, optimized with innovation from Avantor®. Our new brand symbolizes agility and our ability to adapt to customer needs.

Learn More Start Shopping