Tissue processor, ASP300 S
Leverancier: Leica
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Leica ASP300 S
720-2259EA 56870 EUR
720-2259
Tissue processor, ASP300 S
Systemen voor weefselbehandeling
The ASP300 S is a freestanding, fully enclosed vacuum tissue processor for programmable processing of histological tissue specimens. The instrument features three pressure/vacuum options as well as the further option of processing under ambient conditions. Processing temperatures are programmable for each step: 10 reagent stations, three paraffin wax baths, three cleaning stations plus an optional connection for an external cleaning station. The 4,5 litre processing retort is made of electropolished stainless steel for easy cleaning with a contact-free optical sensor to check the fluid level. Retort capacity for up to 300 standard cassettes when using metal baskets or 252 standard cassettes when using the microwave resistant basket set for ASP300 S (optional accessory). An easy-to-use remote reagent handling system can drain and refill reagent bottles from bulk containers via the retort and a hose plugged into the processor module. The three-step retort draining system is tailored to minimise reagent carry-over. A multi-language touch screen user interface combined with a graphical programme display allows even junior operators to quickly become familiar with the system. Ease of use is supported further by direct-access on-line help covering each individual processing step, RemoteCare™.
  • Select your favorite programmes with ease
  • High-quality specimens, every time
  • Full process control
  • Comprehensive specimen protection
Leveringsinformatie: Contains: Basket hook, 3 specimen baskets, stirrer bar, reagent bottle with cap, plastic, 2 set labels, funnel, 2 activated carbon filters, remote fill/drain hose, paraffin drain hose, paraffin wax scraper, plastic, lubricant for valves and O-rings, jumper cable – mains, maintenance kit: (2 spare lids, 9 O-rings), splash protection, allen key, jack connector (remote alarm), 13 reagent bottles with cap, plastic, condensate bottle with cap, plastic, set of power, drip tray, 3,5 Diskette, empty, EC declaration of conformity, multilingual instructions for use ASP300 S.