Portable colorimeters, ion specific, waterproof
Catalog # 705-0685
Supplier: Hanna
Supplier Article Number: HI96704
Local Article Number: HANNHI96704
Discontinued
Specifications
- Model:HI-96704
- Range:0 - 400 µg/l
- Description:Hydrazine meter
- Seq:03
- Automatic switch-off:Yes, after 10 minutes
- Optics:Tungsten lamp with filter and silicon photocell detector
- Power supply:9 V battery
- Weight:0,29 kg
- W×D×H:192×102×67 mm
- Pk:1
Specifications
About this item
Range of microprocessor controlled, handheld, single parameter colorimeters using a monochromatic LED and chemical reagents to promote a colour change within the sample.
- Ideal for portable measurement of environmental water samples
- Accurate and repeatable measured results
- Direct reading on a LCD in g/l or mg/l or µg/l
Ordering information: Reagents and test kits are supplied separately. For full details of the complete range of Hanna portable single parameter colorimeters and reagents please contact your local Avantor office.
Delivery information: Supplied with two cuvettes and two caps, 9 V battery and instruction manual.