Netherlands
Order Entry
Cyclohexane
Cyclohexane
  PROOCHE103
 :  
undefined
Cyclohexane
  PROOCHE103
 :  
  1 * 2 mL
  • Pack type:
    Glass bottle
  • Pk:
    2 mL
  • Seq:
    2145
 :
 :84.16 g/mol
 :13,02752
 :81 °C (1013 hPa)
 :7 °C
 :0.7785 g/cm³ (20 °C)
 :–18 °C
 :Ambient
 :203-806-2
 :1145
 :3,II
 :GHS02
 :GHS08
 :GHS07
 :GHS09
 :110-82-7
 :C₆H₁₂
 :MFCD00003814