Stopcocks, two-way or three-way
Leverancier: Bohlender
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
BOLA
BOHLE654-02EA 117 EUR
BOHLE654-02 BOHLE652-08 BOHLE652-06 BOHLE654-08 BOHLE652-04 BOHLE654-06 BOHLE652-02 BOHLE654-04
Stopcocks, two-way or three-way
Afsluitkranen
Made of PTFE.
  • Temperature resistance upto 110 °C
  • Universal chemical resistance
  • Pressure of 2 bar
Stopcocks for distributing liquids or gases, quick and easy disconnection of flow. The flowing product is only exposed to PTFE
Two-way stopcock with straight bore and two connections. Three-way stopcock with T-shaped bore and three connections, with nuts made of glass-fibre reinforced PTFE for connecting tubing or tube. Conical stopcock plug, tightness is increased by turning the nut on the lower side.