Distilling links, BOLA
Leverancier: Bohlender
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
BOLA
BOHLB303-02EA 306 EUR
BOHLB303-02
Distilling links, BOLA
Onderdelen voor destillatie Destillatieadapters
PTFE, white.
  • Temperature resistance: from −200 to +250 °C
  • Universal chemical resistance
  • Autoclavable
For vertical positioning of the Liebig condenser (e.g. in a lid of a reaction vessel). Integrated special nut (Safe-Lab) for easy locking and unlocking of the ground joint.