Cryogenic trap
Leverancier: SGE (Trajan)
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
SGEA093346EA 1200 EUR
SGEA093346
Cryogenic trap
Cold traps use liquid carbon dioxide injected directly into a special tee fitting surrounding the column. They are suitable for holding components with boiling points equivalent to C7 for 10 to 20 minutes at oven temperatures up to approximately 300 °C.
Cold traps are useful for: On-column trace enrichment, air and gas sampling, purge and trap, multidimensional techniques, head space analysis and splitless injection.
Leveringsinformatie: The kit consists of a cold trap, with pneumatic head switching valve, and all required accessories for installation.