Laboratory notebooks, waterproof
Leverancier: Heathrow Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
HEATHS8610BEA 86.2 EUR
HEATHS8610B HEATHS8610A
Laboratory notebooks, waterproof
Notitieboekjes
For admissibility as court evidence, data on laboratory work, experiment details, and design and development. Use these notebooks to keep your indispensable data. Numbered pages include spaces for title, project number, book number, date, and witness signature.
  • The covers are water proof and chemical resistant
  • Contains periodic and conversion tables in the back