Stoppers, PTFE
Leverancier: Bohlender
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
BOHLH936-10EA 54 EUR
BOHLH936-10 BOHLH937-10 BOHLH936-08 BOHLH936-05 BOHLH937-05 BOHLH936-04 BOHLH936-07 BOHLH937-07 BOHLH936-06 BOHLH936-03 BOHLH937-04 BOHLH936-02
Stoppers, PTFE
Stoppen PE/LDPE/HDPE stoppen
For closing ground joint parts. With ground joint and sealing rings on the outside; knurled or hexagonal grip. Compared with glass stoppers, they are easily removable and can be used without grease.
  • PTFE
  • Temperature resistance from −200 to + 250 °C
  • Chemical resistance
  • FDA conform
The stoppers can expand under heat which might lead to a breaking of the ground joint sockets.