Dipping baskets, SP Bel-Art
Leverancier: Bel-Art Products, a part of SP
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
SCIENCEWARE®
BELAH167160012EA 500 EUR
BELAH167160012
Dipping baskets, SP Bel-Art
Manden Autoclaveerbare manden
Flow through with built-in handles in which liquids flow through the bottoms and sides of these dipping baskets. Made of translucent PP.
  • 3,2 mm thick with 3,2 mm diameter perforations
  • A smooth top rail
  • Solid end plates with 6,4 mm thickness