Barium standard solution, 1,000 mg/L Ba in 0.5 M nitric acid (from Ba(NO₃)₂), standard for AAS
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
BAKR6804.0100EA 71.35 EUR
BAKR6804.0100
Barium standard solution, 1,000 mg/L Ba in 0.5 M nitric acid (from Ba(NO₃)₂), standard for AAS
Certificaten: Traceable to NIST.