Netherlands
Order Entry
Tubing, coloured, PTFE, BOLA
Tubing, coloured, PTFE, BOLA
Supplier:  Bohlender
Certificates
undefined
Tubing, coloured, PTFE, BOLA
Supplier:  Bohlender
Product Information
  • Catalog #
    Supplier #
    Tubing int.Ø
    Wall thickness
    Tubing ext.Ø
    Colour
    Pk
    Availability
    Price
  • BOHLS1862-50
    S1862-50
    6,00 mm
    1,00 mm
    8,00 mm
    Blue
    1 m
    Log in for availability
    Log in for availability
    1 * 1 m
    1 * 1 m - 38,70 €
    38,70 €
    1 * 1 m
       38,70 €
    Specifications:
    Specifications
    Environmentally Preferable
  • BOHLS1862-40
    S1862-40
    4,00 mm
    1,00 mm
    6,00 mm
    Blue
    1 m
    Log in for availability
    Log in for availability
    1 * 1 m
    1 * 1 m - 25,80 €
    25,80 €
    1 * 1 m
       25,80 €
    Specifications:
    Specifications
    Environmentally Preferable
  • BOHLS1861-50
    S1861-50
    6,00 mm
    1,00 mm
    8,00 mm
    Red
    1 m
    Log in for availability
    Log in for availability
    1 * 1 m
    1 * 1 m - 37,30 €
    37,30 €
    1 * 1 m
       37,30 €
    Specifications:
    Specifications
    Environmentally Preferable
  • BOHLS1861-40
    S1861-40
    4,00 mm
    1,00 mm
    6,00 mm
    Red
    1 m
    Log in for availability
    Log in for availability
    1 * 1 m
    1 * 1 m - 25,80 €
    25,80 €
    1 * 1 m
       25,80 €
    Specifications:
    Specifications
    Environmentally Preferable
  • BOHLS1864-50
    S1864-50
    6,00 mm
    1,00 mm
    8,00 mm
    Yellow
    1 m
    Log in for availability
    Log in for availability
    1 * 1 m
    1 * 1 m - 38,00 €
    38,00 €
    1 * 1 m
       38,00 €
    Specifications:
    Specifications
    Environmentally Preferable
  • BOHLS1864-40
    S1864-40
    4,00 mm
    1,00 mm
    6,00 mm
    Yellow
    1 m
    Log in for availability
    Log in for availability
    1 * 1 m
    1 * 1 m - 25,80 €
    25,80 €
    1 * 1 m
       25,80 €
    Specifications:
    Specifications
    Environmentally Preferable
  • BOHLS1863-50
    S1863-50
    6,00 mm
    1,00 mm
    8,00 mm
    Green
    1 m
    Log in for availability
    Log in for availability
    1 * 1 m
    1 * 1 m - 38,00 €
    38,00 €
    1 * 1 m
       38,00 €
    Specifications:
    Specifications
    Environmentally Preferable
  • BOHLS1863-40
    S1863-40
    4,00 mm
    1,00 mm
    6,00 mm
    Green
    1 m
    Log in for availability
    Log in for availability
    1 * 1 m
    1 * 1 m - 25,70 €
    25,70 €
    1 * 1 m
       25,70 €
    Specifications:
    Specifications
    Environmentally Preferable