Netherlands
Order Entry
Ductless laboratory fume hood, benchtop, Basic™ 47
Ductless laboratory fume hood, benchtop, Basic™ 47
Supplier:  LABCONCO
Certificates
undefined
Ductless laboratory fume hood, benchtop, Basic™ 47
Supplier:  LABCONCO
Specifications
  • Catalog No:
  • LBCP2247501
  • LBCP2247301
  • LBCP2246601
  • LBCP2246602
  • Model:
  • Basic™ 47 laboratory hood
  • Basic™ 47 laboratory hood
  • Basic™ 47 laboratory hood
  • Basic™ 47 laboratory hood
  • Lighting:
  • Vapour-proof incandescent lamp
  • Vapour-proof incandescent lamp
  • T8 fluorescent
  • T8 fluorescent
  • Electrical:
  • 220 V, 50 Hz
  • 220 V, 50 Hz
  • 220 V, 50 Hz
  • 220 V, 50 Hz
Specifications
Product Information
  • Catalog #
    Supplier #
    W×D×H ext.
    Weight
    Pk
    Availability
    Price
  • Ductless laboratory fume hood, Basic ™ 47, benchtop, unassembled, for use with remote blower
    Ductless laboratory fume hood, Basic ™ 47, benchtop, unassembled, for use with remote blower
    LBCP2246601
    2246601
    1193,8×635×1219,2 mm
    123 kg
    1
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
    Supplier Article Number: 2246601
    Local Article Number: LBCP2246601
    Specifications:
    More
    Specifications
    Environmentally Preferable
    Model
    Basic™ 47 laboratory hood
    Lighting
    T8 fluorescent
    Electrical
    220 V, 50 Hz
  • Ductless laboratory fume hood, Basic ™ 47, benchtop, unassembled, for use with remote blower
    Ductless laboratory fume hood, Basic ™ 47, benchtop, unassembled, for use with remote blower
    LBCP2246602
    2246602
    1193,8×635×1219,2 mm
    123 kg
    1
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
    Supplier Article Number: 2246602
    Local Article Number: LBCP2246602
    Specifications:
    More
    Specifications
    Environmentally Preferable
    Model
    Basic™ 47 laboratory hood
    Lighting
    T8 fluorescent
    Electrical
    220 V, 50 Hz
  • Ductless laboratory fume hood, Basic ™ 47, benchtop, with integral blower
    Ductless laboratory fume hood, Basic ™ 47, benchtop, with integral blower
    LBCP2247301
    2247301
    1193,8×635×1346,2 mm
    168 kg
    1
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
    Supplier Article Number: 2247301
    Local Article Number: LBCP2247301
    Specifications:
    More
    Specifications
    Environmentally Preferable
    Model
    Basic™ 47 laboratory hood
    Lighting
    Vapour-proof incandescent lamp
    Electrical
    220 V, 50 Hz
  • Basic™ 47 hoods with motor and blower, explosion proof
    Basic™ 47 hoods with motor and blower, explosion proof
    LBCP2247501
    2247501
    1193,8×635×1346,2 mm
    168 kg
    1
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
    Supplier Article Number: 2247501
    Local Article Number: LBCP2247501
    Specifications:
    More
    Specifications
    Environmentally Preferable
    Model
    Basic™ 47 laboratory hood
    Lighting
    Vapour-proof incandescent lamp
    Electrical
    220 V, 50 Hz