Netherlands
Order Entry
Bags, self adhesive, Cleanware™, SP Bel-Art
Bags, self adhesive, Cleanware™, SP Bel-Art
Certificates
undefined
Bags, self adhesive, Cleanware™, SP Bel-Art
Product Information
  • Catalog #
    Supplier #
    Material
    Packed
    L×W
    Thickness
    Pk
    Availability
    Price
  • Disposable waste bag, holds 1,36 kg
    Disposable waste bag, holds 1,36 kg
    129-4930
    F131741008
    HDPE
    50 bags/box
    254×203 mm
    0,025 mm
    50
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST - 51,80 €
    51,80 €
    1 * 50 ST
       51,80 €
    Supplier Article Number: F131741008
    Local Article Number: BELAF131741008
    Specifications:
    Specifications
    Environmentally Preferable
  • Disposable waste bag, holds 1,36 kg
    Disposable waste bag, holds 1,36 kg
    129-4931
    F131741012
    HDPE
    50 bags/box
    406×305 mm
    0,025 mm
    50
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST - 86,40 €
    86,40 €
    1 * 50 ST
       86,40 €
    Supplier Article Number: F131741012
    Local Article Number: BELAF131741012
    Specifications:
    Specifications
    Environmentally Preferable
Frequently Bought Together