Alcohol burners
Leverancier: DWK Life Sciences
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
WHEA237070EA 452 EUR
WHEA237070
Alcohol burners
Verwarmers Alcohollampen
Alcohol burner manufactured from soda-lime glass, designed for use with isopropyl or denatured ethyl alcohol.
  • Low-heat flame is well suited for microscopy purposes
  • With a ground glass stopper
  • Glass cap reduces evoporation of alcohol when not in use