Temperature loggers, Thermo Button
- Catalog No:
- 620-1853
- 620-3186
- 620-3177
- 620-3178
- 620-3183
- 620-3184
- 620-3182
- 620-3179
- 620-3180
- Interface:
- USB
- Battery life:
- 10 years max.
- 30 to 50 cycles, sterilisation process
- 10 years max.
- Data memory - No. of readings:
- 8000
- 8000
- 2000
- RangeAccuracyResolutionPk
- Compact logger, thermo-button, 21GCompact logger, thermo-button, 21G620-3182DAL0052T: –40...+85 °CT: ±1,0 °C0,5 °C11 * 1 ST114,00 €: DAL0052: PRGPDAL0052
Specifications
Environmentally PreferableInterfaceBattery life10 years max.Data memory - No. of readings2000 - Compact logger, thermo- button, 22LCompact logger, thermo- button, 22L620-3183DAL0075T: –40...+85 °CT: ±0,5 °C0,1 ou 0,5 °C11 * 1 ST214,00 €: DAL0075: PRGPDAL0075
Specifications
Environmentally PreferableInterfaceBattery life10 years max.Data memory - No. of readings8000 - Compact logger, thermo-button, 22TCompact logger, thermo-button, 22T620-3184DAL0076T: 0...125 °CT: ±0,5 °C0,1 ou 0,5 °C11 * 1 ST385,00 €: DAL0076: PRGPDAL0076
Specifications
Environmentally PreferableInterfaceBattery life30 to 50 cycles, sterilisation processData memory - No. of readings8000
Software
- RangeAccuracyResolutionPk
- Thermo Track software and USB Thermo Button reader, FR 21 Part 11Thermo Track software and USB Thermo Button reader, FR 21 Part 11620-3186PAC00101 KIT1 * 1 Kit1 480,00 €: PAC0010: PRGPPAC0010
Specifications
Environmentally PreferableInterfaceUSBBattery lifeData memory - No. of readings
Capsule
- RangeAccuracyResolutionPk
- Protection capsule for use in water or for sterilisation processProtection capsule for use in water or for sterilisation process620-1853DAL009611 * 1 ST156,00 €: DAL0096: PRGPDAL0096
Specifications
Environmentally PreferableInterfaceBattery lifeData memory - No. of readings
Holders
- RangeAccuracyResolutionPk
- Key holder, blue: DAL0018: PRGPDAL0018
Specifications
Environmentally PreferableInterfaceBattery lifeData memory - No. of readings - Key holder, green: DAL0020: PRGPDAL0020
Specifications
Environmentally PreferableInterfaceBattery lifeData memory - No. of readings - Key holder, yellow: DAL0021: PRGPDAL0021
Specifications
Environmentally PreferableInterfaceBattery lifeData memory - No. of readings - Key holder, red: DAL0022: PRGPDAL0022
Specifications
Environmentally PreferableInterfaceBattery lifeData memory - No. of readings