Wegwerpsnijplanken, DispoCut™
Leverancier: CellPath
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
DispoCut™
720-2329EA 148 EUR
720-2329 720-2330 720-2328
Wegwerpsnijplanken, DispoCut™
Borden voor dissectie
DispoCut™ is a disposable cutting board designed to increase safety and hygiene in high-risk areas as well we routine cutting areas.
Each DispoCut™ board has a metric and imperial scale to assist in the macroscopic and photographic recording of specimens.