Netherlands
Order Entry
VWR® Silicone Fluids
VWR® Silicone Fluids
undefined
VWR® Silicone Fluids
  •  
    Temperature range
    Viscosity
    Colour
    Pack type
    Pk
  • Silicone fluid 20 cS
    Silicone fluid 20 cS
    630057W
    −40...+200 °C
    20 mm²/s
    Clear
    Plastic container
    20 kg
    1 * 20 kg
    1 * 20 kg - 6 550,00 €
    6 550,00 €
    1 * 20 kg
       6 550,00 €
     : VWRC630057W
  • Silicone fluid 100 cS
    Silicone fluid 100 cS
    630076C
    −40...+200 °C
    100 mm²/s
    Clear
    Plastic bottle
    5 kg
    1 * 5 kg
    1 * 5 kg - 1 440,00 €
    1 440,00 €
    1 * 5 kg
       1 440,00 €
     : VWRC630076C
  • Silicone fluid 100 cS
    Silicone fluid 100 cS
    630077D
    −40...+200 °C
    100 mm²/s
    Clear
    25 kg
    1 * 25 kg
    1 * 25 kg - 4 670,00 €
    4 670,00 €
    1 * 25 kg
       4 670,00 €
     : VWRC630077D
  • Silicone fluid 200 cS
    Silicone fluid 200 cS
    Q1-200KG
    −40...+200 °C
    -
    200 kg
    1 * 200 kg
    1 * 200 kg
     : VWRCQ1-200KG
  • Silicone fluid 350 cS
    Silicone fluid 350 cS
    630094E
    −40...+200 °C
    350 mm²/s
    Clear
    Plastic bottle
    500 g
    1 * 500 g
    1 * 500 g - 192,00 €
    192,00 €
    1 * 500 g
       192,00 €
     : VWRC630094E
  • Silicone fluid 350 cS
    Silicone fluid 350 cS
    630096G
    −40...+200 °C
    350 mm²/s
    Clear
    Plastic bottle
    5 kg
    1 * 5 kg
    1 * 5 kg - 1 520,00 €
    1 520,00 €
    1 * 5 kg
       1 520,00 €
     : VWRC630096G
  • Silicone fluid 350 cS
    Silicone fluid 350 cS
    630097H
    −40...+200 °C
    350 mm²/s
    Clear
    Plastic container
    25 kg
    1 * 25 kg
    1 * 25 kg - 2 530,00 €
    2 530,00 €
    1 * 25 kg
       2 530,00 €
     : VWRC630097H