Sponge, Cleanware™, SP Bel-Art
Leverancier: Bel-Art
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
BELAF170850000EA 52 EUR
BELAF170850000
Sponge, Cleanware™, SP Bel-Art
Schoonmaakdoekjes Sponzen
This steel wool sponge is designed to scrub glassware and stainless steel products sparkling clean with or without detergents.
  • No sharp edges to damage surfaces or irritate hands
  • Pliant sponges are flexible enough to thoroughly clean hard to reach areas
  • Measures roughly 6,4×10 cm
  • Grade 430 steel wool