U heeft gezocht op: Magnetische scheidingsrekken
7  results were found
SearchResultCount:"7"
Sort Results
List view Easy View (new)
Rate These Search Results
Leverancier: Corning
Omschrijving: De magnetische scheidingsapparaten (MSD) zijn essentieel in elk zuiveringsproces op basis van paramagnetische beads. Deze IMAGâ„¢ apparaten zijn ontworp...
Catalogus nummer: (PIERMR02)
Leverancier: Thermo Fisher Scientific
Omschrijving: This separation rack is a magnetic separation device intended for low-throughput magnetic bead manipulation procedures such as magnetic bead-based nuc...
UOM: 1 * 1 Kit
Leverancier: Bel-Art Products, a Part of SP
Omschrijving: Acrylic, clear.
Catalogus nummer: (28-9864-41)
Leverancier: Cytiva
Omschrijving: MagRack Maxi is a magnetic rack for small-scale protein purification and sample enrichment with magnetic beads used for sample volumes up to 50 ml.
UOM: 1 * 1 ST
Catalogus nummer: (28-9489-64)
Leverancier: Cytiva
Omschrijving: MagRack 6 is a magnetic rack for small-scale protein purification and sample enrichment with magnetic beads. Each rack consists of anodized aluminium ...
UOM: 1 * 1 ST
Catalogus nummer: (OZBIMSR1000)
Leverancier: OZ BIOSCIENCES
Omschrijving: Magnetic separation rack designed for 50, 15 or 1,5 ml tubes. It can hold 12 standard microtubes, two 15 ml* and two 50 ml tubes. The magnetic separat...
UOM: 1 * 1 ST
Market Source Item This is a MarketSource item. Additional charges may apply
Catalogus nummer: (OZBIDM30000)
Leverancier: OZ BIOSCIENCES
Omschrijving: Magnetic isolation device is ideal for coupling reactions, washing procedures, aspiration, pipetting etc. MagID is made from an injection moulded plas...
UOM: 1 * 1 ST
Market Source Item This is a MarketSource item. Additional charges may apply
Bel voor prijs
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
This product is marked as restricted and can only be purchased by approved Shipping Accounts. If you need further assistance, email VWR Regulatory Department at Regulatory_Affairs@vwr.com
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
Dit artikel is geblokkeerd voor uw organisatie. Neem contact op met uw inkoopafdeling voor meer informatie
Het orginele artikel is niet langer beschikbaar. Het alternatief dat wordt getoond is beschikbaar
Product(en) gemarkeerd met dit symbool worden niet meer verkocht cq verkocht tot het einde van de voorraad. Alternatieven zijn mogelijk beschikbaar door te zoeken met het VWR-catalogusnummer dat hierboven wordt vermeld. Voor meer informatie kunt u contact opnemen met customer service: 020-4808410.
1 - 7 of 7
no targeter for Bottom