U heeft gezocht op: Geleidbaarheidsstandaarden
42  results were found
SearchResultCount:"42"
Sort Results
List view Easy View (new)
Rate These Search Results
Leverancier: Merck
Omschrijving: These conductivity sachets are convenient, easy to use and ensure accurate and reliable results both in the field or in the laboratory. The 30 ml...
MSDS Sale
Leverancier: Merck
Omschrijving: Conductivity standard, Supelco®, 100000 μ S/cm, Referentietemperatuur: 25 °C
Leverancier: Merck
Omschrijving: Conductivity standard, 1412 µmhos/cm
Catalogus nummer: (CRM498-100G)
Leverancier: Merck
Omschrijving: pH/Conductivity CRM - clay soil
UOM: 1 * 100 g
Leverancier: VWR Chemicals
Omschrijving: A complete range of certified control standards with values ranging from 1,3 to 500000 Âµs to meet all the requirements for electrochemical analys...
Leverancier: VWR Chemicals
Omschrijving: Avantor offers a comprehensive range of conductivity standards as Certified Reference Materials with the primary use calibrating of conductivity meter...
Leverancier: VWR Chemicals
Omschrijving: KCl 0,01 mol/L, VWR®, 1,41 mS/cm, Referentietemperatuur: 25 °C
Catalogus nummer: (85645.260)
Leverancier: VWR Chemicals
Omschrijving: Directly traceable to NIST, tested and certified to ISO17025 and ISO 17034
UOM: 1 * 500 mL
Leverancier: WTW
Omschrijving: Proefset 6R/SET/LAB1 voor geleidbaarheidmeter
Leverancier: Hach
Omschrijving: Sodium chloride standard solutions according to OIML, traceable to NIST and Standard solution verified by Reference Materials Laboratory.
Leverancier: Hach
Omschrijving: The sodium chloride standard solutions are used for calibrating conductivity meters and also used for accuracy checks.
Leverancier: Hach
Omschrijving: For calibrations/accuracy checks on conductivity/salinity meters.
Leverancier: Hach
Omschrijving: Natural Water Standard Solution:40% sodium sulphate/40% sodium bicarbonate/20% sodium chloride.
Leverancier: Hach
Omschrijving: The potassium chloride standard solutions are used for calibrating conductivity cells.
Leverancier: Hach
Omschrijving: Concentratie: 0,1 mol/l KCl, 12 880  µS/cm
Leverancier: Hach
Omschrijving: Potassium chloride standard solutions according to OIML, traceable to NIST and Standard solution verified by Reference Materials Laboratory.
Bel voor prijs
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
This product is marked as restricted and can only be purchased by approved Shipping Accounts. If you need further assistance, email VWR Regulatory Department at Regulatory_Affairs@vwr.com
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
Dit artikel is geblokkeerd voor uw organisatie. Neem contact op met uw inkoopafdeling voor meer informatie
Het orginele artikel is niet langer beschikbaar. Het alternatief dat wordt getoond is beschikbaar
Product(en) gemarkeerd met dit symbool worden niet meer verkocht cq verkocht tot het einde van de voorraad. Alternatieven zijn mogelijk beschikbaar door te zoeken met het VWR-catalogusnummer dat hierboven wordt vermeld. Voor meer informatie kunt u contact opnemen met customer service: 020-4808410.
1 - 16 of 42
no targeter for Bottom